Would you like to log in again?
NEB®5-alpha F Iq Competent E.coli (High Efficiency), New England Biolabs
Supplier: New England Biolabs
C2992H C2992I C2992I C2992H
101417-904EA 176 USD InStock
CA200067-178 101417-904 200067-178 CA101417-904
NEB®5-alpha F Iq Competent E.coli (High Efficiency), New England Biolabs
Competent Cells
An F´ E.coli strain with extremely high transformation efficiency suitable for toxic gene cloning.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty