Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
T7 Express lysY/Iq Competent E.coli (High Efficiency), New England Biolabs
Supplier: New England Biolabs
C3013I C3013I
101417-914EA 200 USD InStock
101417-914 CA101417-914
T7 Express lysY/Iq Competent E.coli (High Efficiency), New England Biolabs
Competent Cells
Enhanced BL21 E.coli derivative, Chemically competent E. coli cells suitable for high efficiency transformation and with the highest level of T7 expression control.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty