Would you like to log in again?
NEB®Express Iq Competent E.coli (High Efficiency), New England Biolabs
Supplier: New England Biolabs
C3037I C3037I
101417-930EA 200 USD InStock
CA101417-930 101417-930
NEB®Express Iq Competent E.coli (High Efficiency), New England Biolabs
Competent Cells
E.coli cells featuring control of IPTG induced expression with non-T7 plasmids.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty