Would you like to log in again?
QUICK BLUNTING™ and QUICK LIGATION™ Kits, New England Biolabs
Supplier: New England Biolabs
E0542S E0542S
101417-934EA 182 USD InStock
CA101417-934 101417-934
QUICK BLUNTING™ and QUICK LIGATION™ Kits, New England Biolabs
Nucleic Acid Reagents Cloning Kits and Ligation
NEB offers the Quick Blunting™ and Quick Ligation™ Kits together as a bundle at a value price.
The Quick Blunting™ Kit is used to convert DNA with incompatible 5´or 3´overhangs to 5´phosphorylated, blunt-ended DNA for efficient blunt-end ligation into DNA cloning vectors. DNA is blunted using T4 DNA polymerase which has both 3´→ 5´ exonuclease activity and 5´→ 3´ polymerase activity. T4 Polynucleotide Kinase is included in the enzyme mix for phosphorylation of the 5´ends of blunt-ended DNA for subsequent ligation into a cloning vector. This kit is optimized for blunting up to 5 µg of DNA in a single reaction. The Quick Ligation™ Kit enables ligation of cohesive end or blunt end DNA fragments in 5 minutes at room temperature (25°C).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty