Would you like to log in again?
QUICK BLUNTING™ Kit, New England Biolabs
Supplier: New England Biolabs
E1201S E1201L E1201S E1201L
101417-942EA 96 USD InStock
CA101417-940 101417-942 101417-940 CA101417-942
QUICK BLUNTING™ Kit, New England Biolabs
Nucleic Acid Reagents Cloning Kits and Ligation
The Quick Blunting Kit is used to convert DNA with incompatible 5´ or 3´ overhangs to 5´ phosphorylated, blunt-ended DNA for efficient blunt-end ligation into DNA cloning vectors
DNA is blunted using T4 DNA polymerase which has both 3´ → 5´ exonuclease activity and 5´ → 3´ polymerase activity. T4 Polynucleotide Kinase is included in the enzyme mix for phosphorylation of the 5´ ends of blunt-ended DNA for subsequent ligation into a cloning vector. This kit is optimized for blunting up to 5 µg of DNA in a single reaction.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty