Would you like to log in again?
Glove Belt Clips, Glove Guard, LP
Supplier: Glove Guard
1939RD 1939YW 1939HVYW 1939PR 1939PK 1939OG 1939LG 1939BK 1939MD 1939GN 1939BL 1939OR 1939GT
10142-012EA 11.31 USD InStock
10142-012 10142-034 10142-024 10142-014 10142-026 10142-030 10142-020 10142-032 10142-022 10142-016 10142-214 10142-028 10142-018
Glove Belt Clips, Glove Guard, LP
Clamps General Purpose Clamps
The Glove Guard® clip is the original glove clip that features a flexible 15 lbs. redundant safety breakaway.
  • Made from an engineered grade material
  • Holds its memory
  • Resists flex-fatigue
  • Non-conductive
  • Non-corrosive
The strong, interlocking teeth of both the small and large ends provide a superior grip to clothing and attached items. These clip is used to attach the small end to belt loop or directly to clothing, and clip the large end to gloves.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty