Would you like to log in again?
Citrajet® Low-Foaming Liquid Acid Cleaners
Supplier: Alconox
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Citrajet®
2015 2015 2001 2005 2001 2055 2055
47732-994EA 1599.87 USD InStock
47732-994 CA47732-992 47732-992 CA47732-996 47732-996 CA47732-994 10142-094
Citrajet® Low-Foaming Liquid Acid Cleaners
Cleaning Agents Detergents and Surface Cleansers
Citrajet is a low foaming, phosphate-free, concentrated, liquid cleaner and metal brightener for high performance removal of metal oxides, scales, salts and inorganic residues in labware washers, parts washers, power-spray systems and manual and ultrasonic cleaning. Corrosion inhibited and free rinsing leaving no interfering residues. Ideal for acid washing and rinsing.
  • Dilute 2:100
  • Ideal for use as acid wash or rinse
  • Concentrated, phosphate free, biodegradable and readily disposable
  • Combines acid and detergent ingredients to remove metal oxides, salts and inorganic residues
Low foaming acid cleaner and rinse for high performance results from your cleaning machinery, such as ultrasonic cleaners.
It can be used to clean laboratory ware, sampling apparatus, dairy equipment optical parts, electronic components, pharmaceutical apparatus, cosmetics manufacturing equipment, metal forgings, stampings and castings, industrial parts, pipes, tanks, boilers and reactors.
And it can be used to remove scale, metal oxides, metal complexes, trace inorganics, milkstone, soil, grit, buffing compound, grime, grease, fats, oils, particulates, amphoteric peptides, deposits, chemical and solvents.
Corrosion inhibited formulation recommended for glass, metal, stainless steel, porcelain, ceramic, plastic, rubber and fiberglass. Can be used on soft metals such as copper, bronze, aluminum, zinc and magnesium if rinsed promptly. Corrosion testing may be advisable.
Cleaning method: Laboratory ware washers, dishwashers, washer-sterilizers, washer-sanitizers, cart washers, spray washers, electro cleaning, ultrasonic cleaners flow through clean-in-place and manual cleaning if proper skin and eye protection is used.
Certifications: USDA compliant. FDA certified.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty