Would you like to log in again?
Regan-Lowe Agar, Hardy Diagnostics
Supplier: Hardy Diagnostics
A65
10142-316PK 157.07 USD InStock
10142-316
Regan-Lowe Agar, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Charcoal Blood Agar with 10% Horse Blood and Cephalexin is a selective medium recommended for the isolation and cultivation of Bordetella pertussis.
The basal medium of Regan-Lowe Agar consists of charcoal agar supplemented with defibrinated horse blood. Charcoal, along with starch, neutralizes fatty acids and peroxides, which are toxic to Bordetella. Horse blood is an added enrichment which supports the growth of Bordetella spp. Cephalexin inhibits the growth of normal flora of the nasopharynx. Yeasts and fungi are inhibited by the inclusion of amphotericin B. Beef extract and enzymatic digest are incorporated in the medium to supply amino acids and other nitrogenous substances that are necessary for bacterial growth. Osmotic equilibrium is maintained by the addition of sodium chloride. Niacin (nicotinic acid) is a vitamin which is added for growth promotion.
Certifications: ATCC No.9797
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty