Would you like to log in again?
SystemSURE Plus™ Luminometer, ATP Hygiene Monitoring System, Hardy Diagnostics
Supplier: Hardy Diagnostics
Ultrasnap™ SystemSURE Plus™
US2020 SS3 US2020 SS3
89492-626EA 2892.26 USD InStock
89492-626 CA89492-626 10142-318 CA10142-318
SystemSURE Plus™ Luminometer, ATP Hygiene Monitoring System, Hardy Diagnostics
Monitoring Systems Hygiene Monitoring Systems
SystemSURE Plus™ is the next-generation system of the SystemSURE II ATP hygiene monitoring system. With improved functionality, new software, and increased memory, the SystemSURE Plus meets the demands of the largest and smallest companies around the world.
  • Provides real-time feedback and facilitates long-term data analysis
  • Optimizes and verifies that cleaning procedures are working
  • Assists in developing and improving process and risk assessment programs (ie. HACCP & GMPs)
  • No scheduled yearly maintenance is necessary
  • Powered by two AA batteries
The SystemSURE Plus™ is designed for convenient, single-hand use to make using the system and navigating the menus easy for every user. Don't let the petite shape fool you - SystemSURE Plus™ makes no compromises when it comes to durability. The rugged outer shell and splash-proof keypad keep SystemSURE Plus safe in the harshest environments. For extra protection, every system comes with a complimentary carrying case with a shoulder strap for hands-free transportation to test locations in your facility.
A clean production environment is a key component to producing safe, quality foods. By incorporating ATP sanitation monitoring into preoperative equipment inspection and change-over cleaning verification, quality personnel can verify equipment cleanliness in seconds. Your facility is only as clean as you start, so it's best to start the day right.
Ordering information: UltraSnap™ Surface ATP Swabs are required for operation but need to be ordered separately.
Delivery information: Kit includes CD software to download data to PC into EXCEL spreadsheet, RS232 cable, operation manual, and durable carrying case.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty