Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pierceable Plug Stopper - For 13 mm Tubes, Globe Scientific

Supplier: Globe Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

113150Y 113150G 113150R 113150W 113150B
10142-474PK 269.55 USD InStock
10142-474 10142-466 10142-470 10142-472 10142-468
Pierceable Plug Stopper - For 13 mm Tubes, Globe Scientific
Stoppers Rubber Stoppers
These pierceable plug stoppers are intended for recapping vacuum blood tubes and test tubes.

  • Thermoplastic elastomer (TPE)
  • Assorted colors available
  • Fits both glass and plastic tubes

The caps are produced from thermoplastic elastomer (TPE) which can easily be pierced with a syringe.

Ordering information: Stoppers are packaged 1000 per bag.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty