Would you like to log in again?
Collection Tube without Cap, 2 ml, IBI Scientific
Supplier: IBI Scientific
IB47086 IB47085
10142-530PK 92.5 USD InStock
10142-530 10142-528
Collection Tube without Cap, 2 ml, IBI Scientific
Tubes Test Tubes
The polypropylene 2 ml collection tubes are ideal when additional collection tubes are required for nucleic acid purifications.
  • Supplied without cap
  • Clean but not sterile
IBI Scientific's 2 ml collection tubes can be used with numerous Nucleic Acid Purification kits found on the market today. These tubes do not have a cap and will mate up with IBI purification kit columns, as well as other competitive columns. Ideal for instances when extra collection tubes are required.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty