Would you like to log in again?
EZ-PECT™, Microbiologics
Supplier: Microbiologics
EZ-PECT™
0485-PEC 0484-PEC 0443-PEC 0486-PEC 0483-PEC 0581-PEC 0392-PEC
10142-718EA 861.77 USD InStock
10142-718 10142-714 10142-736 10142-716 10142-720 10142-722 10142-712
EZ-PECT™, Microbiologics
Control reagents
Antimicrobial Effectiveness and Preservative Efficacy Testing are a critical element to the development of safe and effective Cosmetic and Pharmaceutical products
EZ-PEC™ kits include 20 lyophilized pellets of a single enumerated QC microorganism, 10 vials of hydrating fluid (2 mL per vial) and Peel-Off Certificate of Assay. Quantitative testing saves time and reduces labor by delivering a specific number of colony forming units (CFU).
Ready-to-use EZ-PEC™ quantitative QC microorganism preparations are designed to provide the required microorganism challenge concentrations for Antimicrobial Effectiveness Tests and Preservative Efficacy Tests. When processed as directed, EZ-PEC™ delivers a final concentration of 1.0x105 to 1.0x106 CFU per mL of the product being tested.
Ordering information: To receive this product, customers must have a valid ATCC End User Licensing Agreement in place with Microbiologics. Registration for this agreement is only required once, per location during the first order. Instructions for how to register can be downloaded from the product page. Go to the "Related Information" box, click on "Additional Product Details", then "Instructions for End User Registration with Microbiologics".
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC® cultures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty