Would you like to log in again?
Proinsulin Intact Human ELISA, BioVendor
Supplier: BioVendor
RZ193094100
101422-284EA 1015.77 USD InStock
101422-284
Proinsulin Intact Human ELISA, BioVendor
Assays ELISAs
Proinsulin (PI), a polypeptide of 9390 MW (86 amino acids) is synthetized in the β cells of the Islets of Langerhans in the pancreas.
  • Calibration Range: 0-100 pmol/L
  • Limit of Detection: 0.6 pmol/L
  • Serum: 100 uL/well
Proinsulin (PI), a polypeptide of 9390 MW (86 amino acids) is synthetized in the β cells of the Islets of Langerhans in the pancreas. The sequence of proinsulin is highly conserved in mammalian species and is homologous with IGF-1 and IGF-2. This protein is processed to C-peptide and insulin forms. Both are secreted in equimolar amounts into the blood. In normal individuals, proinsulin is present in the circulatioon in very low concentrations (typical basal values 2–6 pmol/l). The level of proinsulin in serum can be a reflection of β cell status and a consequence of disfunction of PI processing and/or secretion. Proinsulin like material is increased in clinical conditions as insulinoma, familial hyperinsulinemia, non-insulin dependent diabetes mellitus.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty