Would you like to log in again?
IGFBP-2 Mouse/Rat ELISA, BioVendor
Supplier: BioVendor
E08
101422-314EA 904.55 USD InStock
101422-314
IGFBP-2 Mouse/Rat ELISA, BioVendor
Assays ELISAs
IGFBP-2 is, after IGFBP-3, the second most frequent IGFBP in the human blood.
  • Calibration Range: 0.031–2 ng/mL
  • Limit of Detection: 0.01 ng/mL
  • Serum: 10 uL/well
Insulin-like growth factors (IGFs) regulate the proliferation, differentation, apoptose, cell adhesion and metabolism in various tissues and cell types. The IGF-1, which is produced mainly in liver under the influence of growth hormone (GH), regulates as hormone the linear growth of the bones and the process of sexual maturity, while IGF-2 is mainly a growth factor of fetal tissue. The biological actions of IGF over the IGF-Type-1 receptor are modulated variably through the IGF binding proteins (IGFBP-1 to-6). IGFBP-2 is, after IGFBP-3, the second most frequent IGFBP in the human blood. IGFs, especially tumor typical pro-IGF-forms and hormones regulate the expression of IGFBP-2, GH effect is thereby inhibiting. At cellular level IGFBP-2 seems to stimulate the proliferation and dissemination of solid tumors via an IGF-independent mechanism.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty