Would you like to log in again?
IGF-1 Human ELISA, BioVendor
Supplier: BioVendor
E20
101422-316EA 915.29 USD InStock
101422-316
IGF-1 Human ELISA, BioVendor
Assays ELISAs
IGF-1 plays a pivotal role in regulating the proliferation, differentiation and specific functions of many cell types.
  • Calibration Range: 1–50 ng/mL
  • Limit of Detection: 0.09 ng/mL
  • Plasma-Citrate, Plasma-EDTA, Plasma-Heparin, Serum: 20 uL/well
IGF-1 (7649 Da) is a polypeptide protein hormone consisting of 70 amino acids. It is produced primilary by the liver as an endocrine hormone as well as target tissues in a paracrine/autocrine fashion. IGF-1 play a pivotal role in regulating the proliferation, differentiation and specific functions of many cell types.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty