Would you like to log in again?
Human Native Low Density Lipoprotein (from Plasma)
Supplier: BioVendor
RP172562
101422-988EA 1000 USD InStock
101422-988
Human Native Low Density Lipoprotein (from Plasma)
Proteins and Peptides
LDL is a low-density lipoprotein that transports cholesterol and triglycerides from the liver to peripheral tissues. LDL (like all lipoproteins) facilitates the movement of fats and cholesterol within the water based solution of the blood stream. Each natural LDL particle contains a single Apo B-100 molecule (apolipoprotein B-100 is a protein with 4536 amino acid residues) that circulates the fatty acids and keeps them soluble in the aqueous environment. Additionally, the LDL core is highly-hydrophobic, consisting of linoleate (a polyunsaturated fatty acid) and about 1500 esterified cholesterol molecules. This core is enclosed by a shell of phospholipids and unesterified cholesterol in addition to a single copy of B-100 large protein (514 kD). Even though the LDL particles are approximately 22 nm in diameter and have a mass of about 3 million Daltons, they have a mass and size distribution since the LDL particles contain a varying number of fatty acids.
Ordering information: For research use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty