Would you like to log in again?
Human Rhu Interferon-Beta 1B (fromE. coli)
Supplier: BioVendor
RP1762340010
101423-126EA 275.38 USD InStock
101423-126
Human Rhu Interferon-Beta 1B (fromE. coli)
Proteins and Peptides
Interferon beta 1b Human Recombinant produced in E.Coli is a single, non-glycosylated mutein (variant form) of human Interferon beta-1b polypeptide chain containing 165 amino acids and having a molecular mass of 18510.86 Dalton.
Ordering information: For research use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty