Would you like to log in again?
8-OHdG Check, Ultrasensitive, Human ELISA, Multispecies specificity, BioVendor
Supplier: BioVendor
213101
101424-292EA 1047.85 USD InStock
101424-292
8-OHdG Check, Ultrasensitive, Human ELISA, Multispecies specificity, BioVendor
Assays ELISAs
8-OHdG is a widely used biomarker for the measurement of endogenous oxidative DNA damage.
  • Calibration Range: 0.125–10 ng/mL
  • Tissue extract, Urine: 50 uL/well
8-OHdG is a widely used biomarker for the measurement of endogenous oxidative DNA damage.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty