Would you like to log in again?
Insulin Human ELISA, Multispecies specificity, BioVendor
Supplier: BioVendor
YK060
101424-308EA 983.58 USD InStock
101424-308
Insulin Human ELISA, Multispecies specificity, BioVendor
Assays ELISAs
Insulin possesses a wide spectrum of biological actions.
  • Calibration Range: 0.137-100 ng/mL
  • Serum: 25 uL/well
Insulin, a polypeptide hormone with a molecular weight of 5800 Da, is secreted by the beta cells of the islets of Langerhans from the pancreas. Insulin possesses a wide spectrum of biological actions. It stimulates cellular glucose uptake, glucose oxidation, glycogenesis, lipogenesis, proteogenesis and the formation of DNA and RNA. Insulin plays a key role in the regulation of plasma glucose levels (hepatic output inhibition, stimulation of peripheral glucose utilisation). The resulting hypoglycemic effects of insulin are counterbalanced by hormones with hyperglycemic effects (glucagon, growth hormone, cortisol, epinephrin). Insulin secretion is mainly controlled by the plasma glucose levels : hyperglycemia induces a prompt and important increase in circulating insulin levels. Neural influences, as well as various metabolic and hormonal factors (amino acids, glucagon, gastrointestinal hormone) also participate to the control of insulin secretion. Type I (insulin dependent : “juvenile”) diabetes is due to a destruction of the beta cells, with a consequence of absolute lack of insulin. In type II (noninsulin-dependent : “maturity onset”) diabetes, insulin resistance may play an important role; however after several years of evolution, beta-cells failure may occur, leading to a relative insulinopenia requiring, in some cases, insulin administration. Insulin resistance is associated with high circulation levels of the hormone. The most common case of insulin resistance is represented by obesity. Various endocrinopathies (acromegaly, Cushing syndrome) as well as rare cases of insulin receptor defects or cases with anti-insulin receptor antibodies are associated with glucose intolerance or even diabetes due to insulin resistance. The determination of plasma insulin levels is an important parameter in the diagnosis of hypoglycemia.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty