Would you like to log in again?
Human PYY ELISA, Biovendor
Supplier: BioVendor
YK081
101424-312EA 1074.46 USD InStock
101424-312
Human PYY ELISA, Biovendor
Assays ELISAs
96 wells (1 kit) - ELISA is a competitive enzyme immunoassay for the quantitative measurement of PYY.
This EIA kit for determination of PYY in mouse/rat plasma or serum samples is based on a competitive enzyme immunoassay using the combination of antibody to mouse/rat PYY and biotin–avidin affinity system. The 96 wells plate is coated with goat anti rabbit IgG, to which biotin labeled antigen, standard antigen or samples and rabbit anti mouse/rat PYY antibody are added for competitive immunoreaction. After incubation and plate washing, horse radish peroxidase (HRP) labeled streptavidin (SA) is added to form HRP labeled SA-biotinylated antigen-antibody complex on the surface of the wells. Finally, HRP enzyme activity is determined by 3, 3’, 5, 5’-Tetramethylbenzidine (TMB) and the concentration of mouse/rat PYY is calculated.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Caution: This is intended for research use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty