Would you like to log in again?
PYY Human ELISA, BioVendor
Supplier: BioVendor
YK080
101424-314EA 1074.46 USD InStock
101424-314
PYY Human ELISA, BioVendor
Assays ELISAs
PYY shows an inhibitory action on contraction of the gastrointestinal tract and on secretion of pancreatic and gastric juice.
  • Calibration Range: 0.082–6.667 ng/mL
  • Plasma: 25 uL/well
PYY was isolated initially by Tatemoto et al. (1980) from the extract of pig duodenum and shown to be a polypeptide consisting of 36 amino acids residues. PYY is homologous to pancreatic polypeptide (PP) and neuropeptide Y (NPY). PYY is localized mainly in endocrine cells in the intestine (ileum, colon, and rectum). PYY shows an inhibitory action on contraction of the gastrointestinal tract and on secretion of pancreatic and gastric juice. PYY is released by taking diet. The PYY level decreases after resection of the intestine, possiblly be due to the decrease in number of the endocrine cells secreting PYY.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty