Would you like to log in again?
Glucagon-Like Peptide-2, GLP-2, Mouse ELISA, BioVendor
Supplier: BioVendor
YK142
101424-326EA 1164.29 USD InStock
101424-326
Glucagon-Like Peptide-2, GLP-2, Mouse ELISA, BioVendor
Assays ELISAs
GLP-2 has been found to show intestinal epithelial proliferation.
  • Calibration Range: 0.412–100 ng/mL
  • Plasma-Citrate, Plasma-EDTA, Plasma-Heparin, Serum: 25 uL/well
The proglucagon gene is expressed in both pancreatic A cell and intestinal L cell. Tissue-specific posttranslational processing of proglucagon by the prohormone convertase produced the different proglucagon derived peptides (PGDPs) in both pancreas and intestine. The most notable pancreatic PGDP is glucagon, whereas the L cell produced several structurally related peptides, including GLP-2. Among PGDPs, GLP-2 has been found to show intestinal epithelial proliferation.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty