Would you like to log in again?
C-Peptide Rat ELISA, BioVendor
Supplier: BioVendor
633-07279
101424-342EA 1014.77 USD InStock
101424-342
C-Peptide Rat ELISA, BioVendor
Assays ELISAs
Rat C-peptide 1 and 2 are both single chain peptides composed of 31 amino acids.
  • Calibration Range: 30–3000 pg/mL
  • Limit of Detection: 1.5 pg/mL
  • Cell culture medium, Plasma-Citrate, Plasma-EDTA, Plasma-Heparin, Serum: 10 uL/well
Insulin is first synthesized as a single chain polypeptide, proinsulin, then three disulfide bonds are formed, and finally divided into insulin and C-peptide through enzymatic splitting. Rat C-peptide 1 and 2 are both single chain peptides composed of 31 amino acids. Their homology is 93.5%. C-peptide is secreted together with insulin. The role of C-peptide has been considered to keep the best configuration to form three disulfide bonds, and has no biological activity, however, recent studies revealed that C-peptide can bind, probably, a G-protein-coupling specific receptor present on the surface of endothelial cells, kidney microtubule cells and fibroblasts, resulting in activation of calcium-dependent intracellular signaling, activation of Na+- K+-ATPase, and enhancement of NO synthesis. Administration of C-peptide to DM1 patients enhances blood circulation in the skeletal muscle and skin, and also minimizes kidney glomerular hyperfiltration, decreasing albumin excretion into urine, and also improves nervous function, indicating that C-peptide should be given together with insulin to DM1 patients. Important region to bind receptor has been reported to be C-terminal pentapeptide (27–31).
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty