Would you like to log in again?
Anti-dsDNA Mouse ELISA, BioVendor
Supplier: BioVendor
631-02699
101424-348EA 984.92 USD InStock
101424-348
Anti-dsDNA Mouse ELISA, BioVendor
Assays ELISAs
96 wells (1 kit) - ELISA is a competitive enzyme immunoassay for the quantitative measurement of anti-dsDNA.
  • Calibration Range: 15.6 - 1000 mU/ml
  • Limit of Detection: 0.78 mU/ml
  • Plasma-citrate, plasma-EDTA, serum: 1 µl/well
LBIS Mouse anti-dsDNA ELISA Kit is a sandwich ELISA system for quantitative measurement of mouse anti-dsDNA antibody titer. Introduction - Autoantibodies against DNA are IgG type or IgM type reacting with natural double stranded DNA (dsDNA), single-stranded DNA (ssDNA), or both types. In human SLE (systemic lupus erythematosus), there found anti-dsDNA-IgG with high incidence. In SLE blood anti-dsDNA titer closely related to DNA-anti-DNA complex and low complement level, and serves as an important marker for the activity of the disease.
In Shibayagi’s LBIS Mouse anti-dsDNA ELISA Kit, standards or samples are incubated in dsDNA-coated wells to capture anti-dsDNA antibody. After 2 hours incubation and washing, HRP (horse radish peroxidase)-labeled anti-mouse IgG antibody is added, and incubated for 2 hours together with captured anti-dsDNA antibody. After washing, HRP-complex remaining in wells is reacted with a chromogen(TMB) for 20 minutes, and reaction is stopped by addition of acidic solution, and absorbance of yellow product is measured spectrophotometrically at 450 nm. The absorbance is nearly proportional to anti-mouse dsDNA antibody titer. The standard curve is prepared by plotting absorbance against standard dsDNA concentrations. The concentrations in unknown samples are determined using this standard curve.
Caution: This is intended for research use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty