Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-OVA-IgG1 Mouse ELISA, BioVendor

Supplier: BioVendor
630-07669
101424-358EA 1014.77 USD InStock
101424-358
Anti-OVA-IgG1 Mouse ELISA, BioVendor
Assays ELISAs
96 wells (1 kit) - ELISA is a competitive enzyme immunoassay for the quantitative measurement of Anti-OVA-IgG1.

  • Calibration Range: 1.88-120 U/ml
  • Limit of Detection: 0.094 U/ml
  • Plasma-Citrate, Plasma-EDTA, Plasma-Heparin, Serum: 5 ul/well

LBIS Mouse anti-OVA-IgG1 ELISA Kit is an ELISA system for quantitative measurement of mouse anti-OVA-IgG1 antibody titer. 3. Introduction - IgG is the most abundant immunoglobulin and is the main antibody in the secondary immune response. IgG1, with the heavy chain γ1 and molecular weight of 146 kDa, occupies about 2/3 of total IgG. By limiting antigen to OVA (ovalbumin), and by measuring only IgG1, mouse immune response is simplified for elucidation. Shibayagi’s OVA-IgG1 ELISA Kit is useful for specific measurement of mouse anti-OVA IgG1 antibody.

In Shibayagi’s LBIS Mouse anti-OVA-IgG1 ELISA Kit, biotinylated anti-mouse IgG1 antibody, standards or samples are incubated in OVA-coated wells to capture OVA-IgG1. After 1 hour incubation and washing, HRP (horse radish peroxidase)-conjugated streptavidin is added, and incubated for 30 minutes together with captured anti-mouse OVA-IgG1 antibody. After washing, HRP-complex remaining in wells is reacted with a chromogen (TMB) for 20 minutes, and reaction is stopped by addition of acidic solution, and absorbance of yellow product is measured spectrophotometrically at 450 nm. The absorbance is nearly proportional to anti-mouse OVA- IgG1 antibody titer. The standard curve is prepared by plotting absorbance against standard OVA-IgG1 concentrations. The concentrations in unknown samples are determined using this standard curve.

Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.

Caution: This is intended for research use only.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty