Would you like to log in again?
Wright-Giemsa stain, blue, VWR® Quick I™ single step for hematology
Supplier: VWR International
Synonyms: Wright-Giemsa Solution
5231B-32 5231B-16
10143-116CS 6188.45 USD InStock
10143-116 10143-106
Wright-Giemsa stain, blue, VWR® Quick I™ single step for hematology
Wright-Giemsa stain (solutions & kits)
VWR® brand hematology stain offerings satify the needs of laboratory professionals following manual, rapid stain or automated protocols.
  • Stain offerings for multiple protocols
  • Individual stains, reagents and packs have been used in the automated market on the Hematek®, Midas®, Sysmex® and Sakura® instruments
Single-step stains are offered in Wright's and Wright Giemsa formulations and use deionized water to complete the staining process. Three step stain offering yields results comparable to Wright Giemsa stain with results available as soon as 15 seconds.
Certifications: Hematology stain products are manufactured using dyes certified by the Biological Stain Commission. All products undergo Quality Control evaluation using fresh blood.
Order Now
Specification Test Results
pH 6.8 to 7.0@25°C
Visible Scan-(700- 400nm) MA @525 nm 190-240
Visible Scan-(700- 400nm) MA @650 nm 390-475
Performance Differential Blood Smear Morphology
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty