Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eosin Y (yellowish) 1% alcoholic solution, VWR® stain for histology, for cytology

Supplier: VWR International
Danger
Synonyms: Tetrabromofluorescein disodium saltEosin Y disodium saltEosin 2252′,4′,5′,7′-Tetrabromofluorescein disodium saltSolvent Red 43Disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxoxanthen-9-yl)benzoateDisodium 2',4',5',7'-tetrabromo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate)Field's Stain B2,4,5,7-Tetrabromofluorescein disodium salt

7006-32 7006-G
10143-130CS 4239.39 USD InStock
10143-130 10143-132
Eosin Y (yellowish) 1% alcoholic solution, VWR® stain for histology, for cytology
Eosin Y (yellowish)
Eosin Y 1% Alcoholic is a cytoplasmic stain that when used in the hematoxylin and eosin staining procedure provides excellent definition of cell components in shades of red and pink.

  • Crisp, clear staining
  • Well-defined cytoplasm
  • ready-to-use

Eosin Y 1% Alcoholic is a cytoplasmic stain that when used in the hematoxylin and eosin staining procedure provides excellent definition of cell components in shades of red and pink. It is an excellent cytoplasmic stain which provides great contrast to the purple nuclei stained with hematoxylin. Intensified Eosin and Eosin-Phloxine have been formulated to provide the user with increased definition of the cytoplasmic components. These innovative modified eosin counterstains are substituted for Eosin Y 1% Alcoholic in the staining procedure. Timing may be varied to achieve the desired staining intensity. FDA approved for in vitro diagnostic use. For Professional Use Only.


Formula: C₂₀H₆Br₄Na₂O₅
MW: 691.86 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005040
CAS Number: 17372-87-1
UN: 1170
ADR: 3,II
IVDLogo
Order Now
Specification Test Results
Absorbance (Visible Scan 700- 400nm) 410 – 430MA @~531 nm
pH@ 25°C 6.85 – 7.05
Filtration 1 micron nominal

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty