Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eosin Y with Phloxine 1% alcoholic solution, VWR® stain for histology, for cytology

Supplier: VWR International
Danger
Synonyms: Eosin Y – Phloxine BEosin 515 – Phloxine

7009-G 7009-32
10143-140CS 2405.85 USD InStock
10143-140 10143-138
Eosin Y with Phloxine 1% alcoholic solution, VWR® stain for histology, for cytology
Eosin Y with Phloxine
Eosin Y 1% Alcoholic is a cytoplasmic stain that when used in the hematoxylin and eosin staining procedure provides excellent definition of cell components in shades of red and pink.

  • Crisp, clear staining
  • Well-defined cytoplasm
  • ready-to-use

Eosin Y 1% Alcoholic is a cytoplasmic stain that when used in the hematoxylin and eosin staining procedure provides excellent definition of cell components in shades of red and pink. It is an excellent cytoplasmic stain which provides great contrast to the purple nuclei stained with hematoxylin. Intensified Eosin and Eosin-Phloxine have been formulated to provide the user with increased definition of the cytoplasmic components. These innovative modified eosin counterstains are substituted for Eosin Y 1% Alcoholic in the staining procedure. Timing may be varied to achieve the desired staining intensity. FDA approved for in vitro diagnostic use. For Professional Use Only.


Storage Temperature: Ambient
UN: 1170
ADR: 3,II
IVDLogo
Order Now
Specification Test Results
Absorbance (Visible Scan 700- 400nm) @525 nm 310-360 MA
Absorbance (Visible Scan 700- 400nm) @ 510nm 175-210 MA
Filtration 1 micron nominal

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty