Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Wright-Giemsa stain, VWR® Quick III™ Solution II for hematology

Supplier: VWR International
Synonyms: Wright-Giemsa Solution

5330-G 5319-16
10143-232CS 4002.21 USD InStock
10143-232 10143-238
Wright-Giemsa stain, VWR® Quick III™ Solution II for hematology
Wright-Giemsa stain (solutions & kits)
VWR® brand hematology stain offerings satify the needs of laboratory professionals following manual, rapid stain or automated protocols.

  • Stain offerings for multiple protocols
  • Individual stains, reagents and packs have been used in the automated market on the Hematek®, Midas®, Sysmex® and Sakura® instruments

Single-step stains are offered in Wright's and Wright Giemsa formulations and use deionized water to complete the staining process. Three step stain offering yields results comparable to Wright Giemsa stain with results available as soon as 15 seconds.

Certifications: Hematology stain products are manufactured using dyes certified by the Biological Stain Commission. All products undergo Quality Control evaluation using fresh blood.


Order Now
Specification Test Results
Quick III Solution II Absorbance 440-460MN@~650nm
Quick III Solution II pH 6.85-6.95@25°C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty