Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
LiChrospher® Sorbents, MilliporeSigma, Merck
Supplier: Merck
LiChrospher®
1.16178.0010 1.19637.0010 1.16129.0010 1.19638.0010 1.19636.0010 1.16152.0010 1.19641.0010 1.19633.0010 1.19637.0010 1.16152.0010 1.16177.0010 1.19636.0010 1.19641.0010 1.16178.0010 1.16177.0010 1.19640.0010 1.19640.0010 1.19633.0010 1.19638.0010 1.16129.0010 1.19632.0010 1.19632.0010
10144-020EA 755.96 USD InStock
CA1.19636.0010 CA1.19640.0010 CA1.16152.0010 10144-020 10144-022 CA1.19641.0010 CA1.19638.0010 CA1.16177.0010 10144-018 CA1.19633.0010 10143-270 CA1.16129.0010 10143-274 10143-272 10143-310 10143-266 CA1.19632.0010 10143-304 10143-308 CA1.19637.0010 CA1.16178.0010 10143-306
LiChrospher® Sorbents, MilliporeSigma, Merck
Chromatography Sorbents HPLC Sorbents
LiChrospher® is the name given to reliable and versatile traditionally produced spherical silica carriers. LiChrospher® silica carriers are available in a number of different modifications. The polar modified phases LiChrospher® CN, LiChrospher® NH₂ and LiChrospher® Diol as well as LiChrospher® Si with no modification are best for normal phase HPLC.
LiChrospher® Si 60 is a versatile HPLC spherical silica sorbent with a pore diameter of 60 Å and a particle size of 5 µm for normal phase chromatography.
LiChrospher® CN has polar and hydrophobic properties, thus can be used as less polar alternative to LiChrospher® Si 60 in normal phase applications or as less hyfrophobic alternative to LiChrospher® RP-8 in reversed phase applications.
LiChrospher® NH₂ posses polar and hydrophobic properties and can be used for normal phase, reversed phase and ion exchange chromatography. Typical applications are the separations of carbohydrates by reversed phase chromatography or the separsation of nucleotides by ion exhange chromatography.
LiChrospher® Diol has polar and hydrophobic properties, thus can be used as less polar alternative to LiChrospher® Si 60 in normal phase applications or as less hydrophobic alternative to LiChrospher® RP-8 in reversed phase applications. LiChrospher® Diol is also suitable for exclusion chromatography.
LiChrospher® RP-18 and LiChrospher® RP-18 endcapped are reliable and versatile traditionally produced spherical silica gel carriers with C-18 reversed phase properties. They are well suited for the separation of neutral, acidic and weak basic compounds.
LiChrospher® RP-select B is a versatile traditionally produced spherical silica gel carrier with reversed phase properties and with the hydrophobicity of a C-8 phase. This carrier is very well suited for the separation of basic compounds, but with still good properties for the determination of neutral and acidic compounds.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty