Synonyms:
Hydroxybrasilin
, DcH-48
, Natural Black I
7002-16
7002-G
10143-612CS
5397.58
USD
InStock
10143-612
10143-610
Hematoxylin solution (Harris), VWR® stain for histology, for cytology mercury-free, acidified
Hematoxylin staining solutions
General purpose nuclear stain, regressive type. Used with hematoxylin and eosin staining.
- Crisp, clear staining
- Well-defined cytoplasm
- ready-to-use
FDA approved for in vitro diagnostic use. For Professional Use Only.