Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hematoxylin solution (Harris), VWR® stain for histology, for cytology mercury-free, acidified

Supplier: VWR International

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Warning
Synonyms: HydroxybrasilinDcH-48Natural Black I

7002-16 7002-G
10143-612CS 5397.58 USD InStock
10143-612 10143-610
Hematoxylin solution (Harris), VWR® stain for histology, for cytology mercury-free, acidified
Hematoxylin staining solutions
General purpose nuclear stain, regressive type. Used with hematoxylin and eosin staining.

  • Crisp, clear staining
  • Well-defined cytoplasm
  • ready-to-use

FDA approved for in vitro diagnostic use. For Professional Use Only.


Formula: C₁₆H₁₄O₆
MW: 302.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00078111
CAS Number: 517-28-2
Order Now
Specification Test Results
Absorbance (Visible Scan 700- 400nm) @~560 nm 170-230 MA

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty