Would you like to log in again?
Hibar® LiChrosorb® and LiChrospher® HPLC Columns, MilliporeSigma
Supplier: MilliporeSigma
Hibar® LiChrosorb® Hibar® LiChrospher®
1.51902.0001 1.50550.0001 1.50333.0001 1.50433.0001 1.50573.1003 1.50377.0001 1.50545.0001 1.50573.0001 1.50388.0001 1.50332.0001 1.50581.0001 1.50432.0001 1.50318.0001 1.50545.1003 1.51907.0001 1.50581.0001 1.50578.0001 1.50549.0001 1.51903.0001 1.50333.0001 1.50575.0001 1.50546.0001 1.50546.1003 1.50578.0001 1.50579.0001 1.50388.0001 1.50579.0001 1.50549.0001 1.50545.0001 1.50580.0001 1.50574.0001 1.51904.0001 1.50582.0001 1.50574.0001 1.50575.1003 1.50318.0001 1.50477.0001 1.50574.1003 1.50316.0001 1.50547.0001 1.50432.0001 1.50582.0001 1.50316.0001 1.50332.0001 1.50547.0001 1.50575.0001 1.50329.0001 1.50477.1003 1.50583.0001 1.50573.0001 1.50548.0001 1.50546.0001 1.50547.1003 1.50583.0001 1.50477.0001 1.50580.0001 1.50329.0001 1.50377.1003 1.50377.0001 1.50550.0001 1.50433.0001 1.50548.0001
48219-347EA 494.89 USD InStock
CA1.50575.0001 CA1.50583.0001 48219-347 CA48219-354 48219-340 48219-342 CA1.50574.0001 10143-860 CA48219-348 CA48219-347 10143-864 10811-828 10143-862 10143-868 10812-200 10143-866 CA1.50582.0001 CA48219-342 CA1.50545.0001 48219-330 CA48219-340 CA1.50548.0001 10811-832 10811-830 10812-190 10143-872 48219-326 48219-328 10143-870 CA1.50550.0001 10143-876 10812-196 10143-874 10812-198 10143-878 CA1.50333.0001 CA1.50547.0001 48219-322 48219-324 CA1.50580.0001 CA48219-328 CA48219-326 CA48219-324 10143-880 CA1.50578.0001 EM1.51903.0001 10812-202 10812-204 CA1.50579.0001 10812-206 10812-208 CA48219-322 48219-354 48219-348 CA1.50546.0001 CA1.50573.0001 10143-854 10143-852 CA1.50549.0001 10143-858 CA1.50581.0001 10143-856
Hibar® LiChrosorb® and LiChrospher® HPLC Columns, MilliporeSigma
Chromatography Columns
For analytical HPLC.
RP-18 is reversed phase C18 (with an octadecylsilyl chain). RP-8 is reversed phase C8 (with an octylsilyl chain).
Cartridge holder 48219-176 is for the direct coupling of Hibar® columns. The holder replaces the original top fitting and holds any 4x4mm LiChroCART® guard column.
LiChrosorb® columns are available with 5µm or 10µm irregular media. LiChrospher® columns are available with 5 or 10 µm spherical media. Stainless steel columns are complete with fittings at both ends and shipped with an individual test chromatogram. May be connected to any HPLC instrument using standard 1.6mm (¹/₁₆") capillary male connections. To prolong the life of analytical columns, use with suitable precolumns.
Ordering information: Hibar® columns include end fittings.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty