Would you like to log in again?
LiChrosorb® Sorbents, MilliporeSigma, Merck
Supplier: Merck
LiChrosorb®
1.09318.0010 1.09332.0010 1.09334.0010 1.09332.0010 1.09333.0010 1.09318.0010 1.09309.0010 1.09333.0010 1.09334.0010 1.09309.0010
10143-784EA 1128.18 USD InStock
CA1.09309.0010 CA1.09318.0010 CA1.09334.0010 CA1.09333.0010 10143-784 10144-012 10143-782 10144-014 CA1.09332.0010 10144-016
LiChrosorb® Sorbents, MilliporeSigma, Merck
Chromatography Sorbents HPLC Sorbents
LiChrosorb® Si 100 is a reliable irregular shaped silica sorbent with a pore diameter of 100 Å for normal phase chromatography.
LiChrosorb® RP-8 is a reliable irregular shaped silica gel carrier with reversed phase properties. It is well suited for the separation of neutral, acidic and weak basic compounds.
LiChrosorb® RP-18 is a reliable irregular shaped silica gel carrier with reversed phase properties. It is well suited for the separation of neutral, acidic and weak basic compounds.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty