Would you like to log in again?
Superspher® Sorbents, MilliporeSigma, Merck
Supplier: Merck
Superspher®
1.19643.0010 1.19643.0010 1.19618.0010 1.19613.0010 1.19613.0010 1.19612.0010 1.19618.0010 1.19612.0010
10143-300EA 2378.76 USD InStock
CA1.19618.0010 CA1.19613.0010 10143-300 CA1.19643.0010 10143-298 10144-026 10143-302 CA1.19612.0010
Superspher® Sorbents, MilliporeSigma, Merck
Chromatography Sorbents HPLC Sorbents
Superspher® is a high performance silica carrier with a particle size of 4 µm which yields an excellent pressure/separation performance ratio.
Superspher® RP-8 is a reliable and versatile traditionally produced spherical silica gel carrier with C-8 reversed phase properties. This carrier is well suited for the separation of neutral, acidic and weak basic compounds.
Superspher® RP-18 and Superspher® RP-18 endcapped are reliable and versatile traditionally produced spherical silica gel carriers with C-18 reversed phase properties. They are well suited for the separation of neutral, acidic and weak basic compounds.
Superspher® RP-select B is a highly reproducible traditionally produced spherical silica gel carrier with reversed phase properties and the hydrophobicity of a C-8 phase. This carrier is very well suited for the separation of basic compounds, but with still good properties for the determination of neutral and acidic compounds.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty