Would you like to log in again?
Micro-organism Lysing VK01, Precellys® Lysing Kit, Bertin
Supplier: Bertin
P000914-LYSK0-A.0 P000929-LYSK0-A.0
10144-562EA 139.96 USD InStock
10144-562 10144-504
Micro-organism Lysing VK01, Precellys® Lysing Kit, Bertin
Beads Glass Beads
VK01 is made with 0.1 mm inert glass beads and designed for micro-organism lysing like gram-, gram+ bacteria, spores and more.
  • Ideal for yeast, bacteria and fungi lysis
  • Dnase/Rnase free
  • Autoclavable
Use with the Precellys®24, Precellys® Evolution, or Minilys homogenizers.
50 tubes (prefilled)
Certifications: Dnase/Rnase free certified.
Delivery information: Packaged 1 kit contain 50 prefilled and ready-to-use tubes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty