Would you like to log in again?
Bacteria Lysing CK01, Precellys® Lysing Kit, Bertin
Supplier: Bertin
Precellys®
P000938-LYSK0-A.0 P000919-LYSK0-A.0 P000932-LYSK0-A.0 P000948-LYSK0-A.0
10144-590EA 253.34 USD InStock
10144-590 10144-510 10144-533 10144-568
Bacteria Lysing CK01, Precellys® Lysing Kit, Bertin
Homogenizers Tissue Grinders
CK01 is made with inert 0.1 mm ceramic (zirconium oxide) beads and designed for micro-organism lysing like gram+, gram- bacteria, spores and more.
  • Ideal for bacteria and spores lysis
  • Autoclavable
  • Usable with Precellys®24, Precellys® Evolution and Minilys® homogenizers
Certifications: Dnase/Rnase free certified.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty