Would you like to log in again?
7 ML DUAL WHITE BLOCKING PLATE
Supplier: BERTIN CORP
Part for Precellys® 24 Dual, allows to process up to 6 tubes of 7mL
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
7 ML DUAL WHITE BLOCKING PLATE 10144-548 S000687-PR241-A.0 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty