Would you like to log in again?
Precellys® CK68 Lysing Kit, Dry Hard Tissue Grinding, Bertin
Supplier: Bertin
P000923-LYSK0-A.0 P000950-LYSK0-A.0 P000931-LYSK0-A.0
10144-592EA 266.83 USD InStock
10144-592 10144-522 10144-566
Precellys® CK68 Lysing Kit, Dry Hard Tissue Grinding, Bertin
Homogenizers Tissue Grinders
CK68-R is made with inert 2.8 mm ceramic (zirconium oxide) beads.
  • Autoclavable
  • Usable with Precellys®24, Precellys® Evolution and Minilys® homogenizers
It is designed for dry samples grinding like rice grains, pepper grains and more.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty