Would you like to log in again?
DAPI Fluoromount-G®, Southern Biotech
Supplier: Southern Biotechnology
Fluoromount™
0100-20
101442-494EA 65.38 USD InStock
101442-494
DAPI Fluoromount-G®, Southern Biotech
Stains Microscopy and staining reagents
DAPI Fluoromount-G® is suitable for mounting frozen and paraffin sections, whole mounts, and cells.
  • For mounting frozen and paraffin sections, whole mounts, and cells requiring nuclear visualization
  • Clear liquid mounting medium
  • Water soluble
  • Semi-permanent seal
  • Applications: IHC-FS, IHC-PS, ICC, (F)ISH, TUNEL, PLA, Migration, Apop
DAPI Fluoromount-G® is a water soluble, 4',6-diamidino-2-phenylindole (DAPI) containing compound recommended for slides mounted after a staining procedure having an aqueous final step. When bound to DNA, DAPI fluoresces and is typically used as a nuclear/chromosomal counterstain. Since DAPI Fluoromount-G® is water soluble, the coverslip may be removed by submerging the slide in a PBS solution until the coverslip is loosened. This mounting medium also provides a semi-permanent seal for storage of slide preparations.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty