Would you like to log in again?
1M Citric acid/ Sodium Hydroxide, Buffer, Rigaku
Supplier: Rigaku
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
1008159 1008161 1008162 1008160
101443-590EA 149.08 USD InStock
101443-590 101443-594 101443-268 101443-270
1M Citric acid/ Sodium Hydroxide, Buffer, Rigaku
Buffers pH Calibration Buffers
Use Rigaku stock solutions directly or as a component in solutions you create in your own lab. Stock solutions are prepared using high quality raw materials, ASTM Type 1 water, and sterile packaging. 
Caution: Intended for Research and Development Use Only.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty