Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

tri-Sodium citrate solution 1.6 M

Supplier: Rigaku Reagents

1008082 1008083
101443-386EA 82.38 USD InStock
101443-386 101443-744
tri-Sodium citrate solution 1.6 M
tri-Sodium citrate solutions
Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.


Formula: Na₃C₆H₅O₇·2H₂O
MW: 294.1 g/mol
MDL Number: MFCD00150031
CAS Number: 6132-04-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty