Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diammonium hydrogen orthophosphate 2 M

Supplier: Rigaku Reagents
Synonyms: Ammonium Phosphatedi-Ammonium hydrogen phosphateDiammonium hydrogen orthophosphateAmmonium hydrogen phosphateAmmonium monohydrogen phosphateAmmonium phosphate dibasic

1008331 1008330
101443-440EA 81.08 USD InStock
101443-440 101443-830
Diammonium hydrogen orthophosphate 2 M
di-Ammonium hydrogen orthophosphate
Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.


Formula: (NH₄)₂HPO₄
MW: 132.06 g/mol
MDL Number: MFCD00010891
CAS Number: 7783-28-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty