Would you like to log in again?
LONGAMP® Taq PCR Kit, New England Biolabs
Supplier: New England Biolabs
LONGAMP®
E5200S E5200S
101444-720EA 135 USD InStock
101444-720 CA101444-720
LONGAMP® Taq PCR Kit, New England Biolabs
Nucleic Acid Reagents End-point PCR Enzymes and Kits
The LongAmp Taq PCR Kit features LongAmp Taq DNA Polymerase, which is a unique blend of Taq and Deep VentR ™ DNA Polymerases
The 3´→5´ exonuclease activity of Deep VentR Polymerase increases the fidelity and robust amplification of Taq DNA Polymerase. LongAmp Taq DNA Polymerase offers two fold higher fidelity than Taq Polymerase alone. The LongAmp Taq PCR Kit is supplied with LongAmp Taq Reaction Buffer, which is optimized to give highest PCR yields up to 30 kb. The kit can generate a wide range of PCR products, up to 30 kb from plasmid or from human genomic DNA.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty