Would you like to log in again?
IMPACT™ Kit for Protein Purification, New England Biolabs
Supplier: New England Biolabs
E6901S E6901S
101444-722EA 495 USD InStock
101444-722 CA101444-722
IMPACT™ Kit for Protein Purification, New England Biolabs
Protein Linking and Labeling Systems
The IMPACT (Intein Mediated Purification with an Affinity Chitin-binding Tag) system is a novel protein purification system which utilizes the inducible self-cleavage activity of protein splicing elements (termed inteins) to separate the target protein from the affinity tag.
It distinguishes itself from all other purification systems by its ability to purify, in a single chromatographic step, a native recombinant protein without the use of a protease. Each intein tag contains a chitin binding domain (CBD) for the affinity purification of the fusion protein on a chitin resin. Induction of on-column cleavage, using thiol reagents such as dithiothreitol (DTT), releases the target protein from the intein tag. The vectors included in this kit allow for the fusion of the target protein at its C-terminus (pTXB1) or at its N-terminus (pTYB21) to the intein tag.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty