Would you like to log in again?
GpC Methyltransferase (M.CviPI), New England Biolabs
Supplier: New England Biolabs
M0227L M0227S M0227L M0227S
101444-740EA 86 USD InStock
CA101444-738 101444-740 CA101444-740 101444-738
GpC Methyltransferase (M.CviPI), New England Biolabs
Enzymes
The GpC Methyltransferase (M.CviPI) methylates all cytosine residues (C5) within the double-stranded dinucleotide recognition sequence 5´...GC...3´.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty