Would you like to log in again?
Oligo dT25 Magnetic Beads, New England Biolabs
Supplier: New England Biolabs
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
S1419S S1419S
101444-826EA 356 USD InStock
CA101444-826 101444-826
Oligo dT25 Magnetic Beads, New England Biolabs
Beads Magnetic Beads
Oligo d(T)25 Magnetic Beads are a 1 μm polymer based affinity matrix for the small-scale isolation of mRNA from crude cell lysates and tissue.
The isolation occurs through the hybridization of covalently coupled oligo d(T)25 to the poly(A) region present in most eukaryotic mRNA. Applications include direct mRNA isolation following lysis and second-round purification of previously isolated total RNA. The magnetic separation technology is scalable and permits elution of intact mRNA in small volumes eliminating the need for precipitation of the isolated mRNA. Beads can be reused up to three times and the researcher has the option of eluting the isolated mRNA or using the bound d(T)25 as a primer in a first-strand cDNA reaction.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty