Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 30% (w/w) in aqueous solution traceable to NIST Standard Reference Material (SRM)

Supplier: AQUA SOLUTIONS MS
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

8606-500ML 8606-4L 8606-4LC 8606-1L 8606-20L
101223-524EA 29.93 USD InStock
101223-524 101226-852 101446-044 101225-730 101226-410
Sodium hydroxide 30% (w/w) in aqueous solution traceable to NIST Standard Reference Material (SRM)
Sodium hydroxide


Formula: NaOH
MW: 40 g/mol
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1824
ADR: 8,II
Order Now
Specification Test Results
Appearance and color Clear water white liquid
Assay NaOH (w/w) 29.70 - 30.30%
Density 1.32305 - 1.33249

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty