Would you like to log in again?
Tryptic Soy Agar (TSA), Teknova
Supplier: Teknova
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
T0520 T0401 T0500 T0502 T0500 T0515
101320-642EA 2391.5 USD InStock
101320-642 CA101320-642 200059-614 200059-616 200059-618 101447-042
Tryptic Soy Agar (TSA), Teknova
Media for Microbiology Dehydrated Media for Microbiology
TSA is a general purpose media produced via enzymatic digestion of soybean meal and casein. TSA is frequently the base media of other agar types; for example, blood agar plates are made by enriching TSA plates with blood.
TSA plates support growth of many semi-fastidious bacteria, including some species of Brucella, Corynebacterium, Listeria, Neisseria, and Vibrio.
1.5% Tryptone, 0.5% Soytone, 0.5% Sodium Chloride, 1.5% Agar.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty