Would you like to log in again?
Alpha 1-3,6 Galactosidase
Supplier: New England Biolabs
P0731S P0731S
101447-570EA 151 USD InStock
CA101447-570 101447-570
Alpha 1-3,6 Galactosidase
Enzymes
α1-3, 6 Galactosidase is a highly specific exoglycosidase that catalyzes the hydrolysis of α1-3, 6 linked D-galactopyranosyl residues from oligosaccharides.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty